Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Brain Whole Tissue Lysate (Post-Natal Whole)
Shop All Novus Biologicals Products

Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Western Blot, ELISA, Immunoprecipitation, SDS-Page |
| Quantity | 200 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | Rat |
| Buffer | 1 x RIPA Buffer with HALT Protease and Phosphatase Inhibitors |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?