Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat CALCB ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP342640 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP342640 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP342640 Supplier R&D Systems™ Supplier No. NBP342640

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Chemiluminescence ELISA Kit

Specifications

Assay Range 7.81 to 2000 pg/mL
Assay Sensitivity 2.67 pg/mL
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Line Novus
Sample Type Serum, Plasma, Tissue Homogenates, Cell Lysates, Cell Culture Supernatant
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 797
Gene Symbol CALCB
Kit Contents Pre-coated 96T Strip Plate, Plate sealer for 96 wells, Standard, Standard Diluent, Detection Reagent A, Assay Diluent A, Detection Reagent B, Assay Diluent B, Substrate A, Substrate B, Wash Buffer (30X concentrate), Instruction manual
Quantity 1 Kit
Research Discipline Cardiovascular Biology, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym calcitonin-related polypeptide beta, FLJ30166
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.