Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat Carboxypeptidase-B Recombinant Protein, PeproTech®

Catalog No. 4000050UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400-00-50UG 50 μg
40000100UG 100 μg
400-00-1MG 1 mg
40000250UG 250 μg
400-00-25UG 25 μg
40000500UG 500 μg
400-00-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 400-00-50UG Supplier Gibco™ Supplier No. 4000050UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19223
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 24271
Molecular Weight (g/mol) 35.1 kDa
Name Rat Carboxypeptidase-B
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); Cpb; Cpb1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Common Name Carboxypeptidase B1
Gene Symbol CPB1
Biological Activity Carboxypeptidase-B sequentially cleaves C terminal K and R residues.
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASGHSYTKYN NWETIEAWIQ QVATDNPDLV TQSVIGTTFE GRNMYVLKIG KTRPNKPAIF IDCGFHAREW ISPAFCQWFV REAVRTYNQE IHMKQLLDEL DFYVLPVVNI DGYVYTWTKD RMWRKTRSTM AGSSCLGVDP NRNFNAGWCE VGASRSPCSE TYCGPAPESE KETKALADFI RNNLSTIKAY LTIHSYSQMM LYPYSYDYKL PENYEELNAL VKGAAKELAT LHGTKYTYGP GATTIYPAAG GSDDWSYDQG IKYSFTFELR DTGFFGFLLP ESQIRQTCEE TMLAVKYIAN YVREHLY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.