Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Ccl2 Recombinant Protein

Catalog No. 89938380 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-380 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-380 Supplier Abnova™ Supplier No. P4848
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: QPDAVNAPLTCCYSFTGKMIPMSRLENYKRITSSRCPKEAVVFVTKLKREICADPNKEWVQKYIRKLDQNQVRSETTVFYKIASTLRTSAPLNVNLTHKSEANASTLFSTTTSSTSVEVTSMTEN

Specifications

Accession Number P14844
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 24770
Molecular Weight (g/mol) 14.1kDa
Name Ccl2 (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue
Quantity 10 μg
Immunogen QPDAVNAPLTCCYSFTGKMIPMSRLENYKRITSSRCPKEAVVFVTKLKREICADPNKEWVQKYIRKLDQNQVRSETTVFYKIASTLRTSAPLNVNLTHKSEANASTLFSTTTSSTSVEVTSMTEN
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias MCP-1/Scya2/Sigje
Common Name Ccl2
Gene Symbol Ccl2
Biological Activity The activity is determined by the ability of MCP-1 to chemoattract THP-1 cells and is detectable starting at 10ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.