Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat CD40 Ligand/TNFSF5 (HA-tag) Protein

Catalog No. 8414CL025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-14C-L025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-14C-L025 Supplier R&D Systems™ Supplier No. 8414CL025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured in a cell proliferation assay using mouse splenic B cells in the presence of IL-4. Banchereau, J. et al. (1991) Science 251:70. The ED50 for this effect is 0.15-0.9 ng/mL in the presence of 0.1 μg/mL of the cross-linking antibody, Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).

Specifications

Accession Number Q9Z2V2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 959
Molecular Weight (g/mol) M.W. (Observed): 22 - 30 kDa, reducing conditions; M.W. (theoretical): 22 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived rat CD40 Ligand/TNFSF5 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Rat CD40 Ligand (Met112-Leu260 (N-terminus) Accession # Q9Z2V2 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Common Name CD40 Ligand/TNFSF5
Gene Symbol Cd40lg
Biological Activity Measured in a cell proliferation assay using mouse splenic B cells in the presence of IL-4. Banchereau, J. et al. (1991) Science 251:70. The ED50 for this effect is typically 0.15-0.9ng/mL in the presence of 0.1μg/mL of the cross-linking antibody, Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody.
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in steril PBS containing (at least 0.1% human or bovine serum albumin.)
Expression System Chinese Hamster Ovary cell line, CHO-derived HA (YPYDVPDYA), N-terminus; GCN4-IZ; GGGSGGGSGGGS; Rat CD40 Ligand (Met112-Leu260 Accession # Q9Z2V2, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE with silver staining.
Protein CD40 Ligand/TNFSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.