Learn More
Sino Biological Rat CD59 MAb 25TESTS 80299-R014-F-25
Supplier:  Sino Biological 80299R014F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD59 (rR CD59; Catalog#80299-R08H; P27274; Met1-Asn100) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-9533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.