Learn More
Sino Biological Rat CD6 MAb 25TESTS 80311-MM01-A-25
Supplier:  Sino Biological 80311MM01A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD6 / Cluster of Differentiation 6 (rR CD6 / Cluster of Differentiation 6; Catalog#80311-R08H; Q812A4; Met1-Asn396) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-2029
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.