Learn More
Specifications
Specifications
| Gene | CD8 beta |
| Accession Number | CAA27850.1 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 640 bp |
| Gene ID (Entrez) | 926 |
| Gene Alias | CD8 antigen, beta polypeptide 1 (p37), CD8 beta, CD8b, CD8b antigen, CD8b molecule, CD8B1, CD8B1P37, LEU2, LY3, LYT3, MGC119115, P37, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rat |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
