Learn More
Biomatik Corporation Rat Cluster Of Differentiation 19 (CD19) ELISA Kit
Supplier:  Biomatik Corporation EKU03230
Encompass_Preferred
Rat Cluster Of Differentiation 19 (CD19) ELISA Kit for detecting CD19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1LNH2. Target Synonyms: B4; B-lymphocyte surface antigen B4; T-cell surface antigen Leu-12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9055
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.