Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat CNTF Recombinant Protein, PeproTech®

Catalog No. 4505050UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-50-50UG 50 μg
45050100UG 100 μg
450-50-1MG 1 mg
45050250UG 250 μg
450-50-25UG 25 μg
45050500UG 500 μg
450-50-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 450-50-50UG Supplier Gibco™ Supplier No. 4505050UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat CNTF is synthesized as a 199 amino acid polypeptide (22.7 kDa) lacking a hydrophobic N-terminal signal for secretion. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20294
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 25707
Molecular Weight (g/mol) 22.7 kDa
Name Rat CNTF
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF
Common Name CNTF
Gene Symbol CNTF
Biological Activity Determined by its ability to stimulate proliferation of human TF-1 cells using a concentration range of 25.0-35.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AFAEQTPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNINLD SVDGVPVAST DRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHT LMLQVSAFAY QLEELMVLLE QKIPENEADG MPATVGDGGL FEKKLWGLKV LQELSQWTVR SIHDLRVISS HQMGISALES HYGAKDKQM
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.