Learn More
American Research Products Inc Rat coagulation factor XIII F
Supplier: American Research Products Inc CSBE10122R
Rat coagulation factor XIII FXIII ELISA Kit from CUSABIO Detection Range 4 7 ng/ml-300 ng/ml Sensitivity 1 175 ng/ml Sample serum plasma tissue homogenates Method Sandwich
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.