Learn More
ACROBiosystems Rat Complement C2 200ug
Supplier:  ACROBiosystems CO2R82E6200UG
Encompass_Preferred
Biotinylated Rat Complement C2 Protein HisAvitag (CO2-R82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 758 (Accession # Q6MG73).
Catalog No. 50-253-5225
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.