Learn More
Biomatik Corporation Rat Complement Component 1, Q Subcomponent A (C1qA) ELISA Kit
Supplier:  Biomatik Corporation EKU03375
Encompass_Preferred
Rat Complement Component 1, Q Subcomponent A (C1qA) ELISA Kit for detecting C1qA in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.67ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31720. Gene ID: 298566. Target Synonyms: Complement Component 1,q Subcomponent,Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.