Learn More
R&D Systems™ Rat Cortical Stem Cells (3 x 10e6 cells/vial)
SDP
Catalog No. NSC001 Shop All Novus Products
Change view
Click to view available options
Quantity:
3 x 10e6 cells/vial
Catalog No. Quantity
NSC001 3 x 10e6 cells/vial
1 options
This item is not returnable. View return policy
Catalog No. NSC001 Supplier R&D Systems™ Supplier No. NSC001
Add to Cart
Add to Cart
This item is not returnable. View return policy
For cortical stem cell expansion and differentiation into neurons, astrocytes and oligodendrocytes
  • Have been fully characterized to reduce experimental variation
  • Can be expanded as monolayers or neurospheres
  • Are free of mycoplasma and microbial contamination
Specifications
Product Line R&D Systems
Quantity 3 x 10e6 cells/vial