Learn More
Gibco™ Rat CXCL1 (GRO alpha) Recombinant Protein, PeproTech®
Description
400-10-1MG will be provided as 2 x 500 μg (400-10-500UG). Recombinant Rat GRO/KC is a 7.8 kDa protein consisting of 72 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P14095 |
| For Use With (Application) | Control, Western Blot Control, ELISA Standard, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 81503 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Name | Rat CXCL1 (GRO alpha) |
| Purification Method | Purified |
| Quantity | 25 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.