Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat CXCL5/ENA-78 ELISA Kit (Colorimetric)
SDP

Catalog No. NB001538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001538 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001538 Supplier Novus Biologicals™ Supplier No. NBP278751
Only null left
Add to Cart
Add to Cart

Specifications

Accession Number P97885
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 6374
Gene Symbol CXCL5
Host Species Rat
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target CXCL5/ENA-78
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.