Learn More
Description
- Standard (18°) Ear Bars taper at the tip to penetrate far into the ear canal for a more secure hold.
- Non-puncture Ear Bars have a 45° taper and do not puncture the tympanic membrane
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| For Use With (Equipment) | 51800 and 51850 series |
| Size | 45° |
| Animal | Rat |
| Model | Non-puncture rat ear bars, 45° |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
