Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat FGF-7 (KGF) Recombinant Protein

Catalog No. PIRP10932
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP10932 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP10932 Supplier Invitrogen™ Supplier No. RP10932
Add to Cart
Add to Cart

Recombinant Protein

This protein binds to keratinocytes and most parenchymal epithelial cells; binds specific splice variant of FGF receptor expressed in epithelial cells (FGFr2IIIb). Dilute with either 0.1% TFA/40% acetonitrile or PBS/0.1% CHAPS for best stability and to prevent absorption of product on walls of vessels.

Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02195
Concentration 0.346 mg/mL
For Use With (Application) Control
Formulation PBS with no preservative
Gene ID (Entrez) 29348
Molecular Weight (g/mol) 17 kDa
Name Rat FGF-7 (KGF)
Quantity 20 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Common Name KGF
Gene Symbol Fgf7
Biological Activity The activity of rat FGF-7 at 1-10 ng/mL is determined by tritiated thymidine incorporation using cultured mouse keratinocytes. Maximum stimulation observed (at 1 ng/mL)
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.