Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat FGF-acidic (FGF-1) Recombinant Protein, PeproTech®

Catalog No. 40029A100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40029A100UG 100 μg
40029A10UG 10 μg
40-029-A01M 2 x 500 μg
40029A250UG 250 μg
40029A500UG 500 μg
40029A50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 40029A100UG Supplier Gibco™ Supplier No. 40029A100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

400-29A-1MG will be provided as 2 x 500 μg (400-29A-500UG). Recombinant Rat FGF-acidic is a 15.9 kDa protein consisting of 141 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61149
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 25317
Molecular Weight (g/mol) 15.9 kDa
Name Rat FGF-acidic (FGF-1)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product
Common Name FGF1
Gene Symbol FGF1
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.1 ng/ml in the presence of 10 ug/ml heparin, corresponding to a specific activity of ≥ 1 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.