Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat GFAP ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349738 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349738 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349738 Supplier Novus Biologicals™ Supplier No. NBP349738
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Rat GFAP in samples. No significant cross-reactivity or interference between Rat GFAP and analogues was observed
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 2670
Gene Symbol GFAP
Quantity 1 Kit
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.