Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Ghrelin (aa 252-267) Synthetic Peptide

Catalog No. ABC290
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC290 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC290 Supplier Invitrogen™ Supplier No. PEP290

Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by the supplier. Please to view product availability in your area.

Synthetic Peptide

Rat Obestatin (residues 252-267) Synthetic Peptide, PEP-290, from Invitrogen.

GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake. PEP-290 is a 23 amino acid synthetic peptide whose sequence is derived from the rat Obestatin. The sequence of this peptide is (amino to carboxy terminus): F-N-A-P-F-D-V-G-I-K-L-S-G-A-Q-Y-Q-Q-H-G-R-A-LThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, Obestatin, catalog &Num; PA1-1070. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 59301
Species Rat
Purification Method purified
Accession Number Q9QYH7
Common Name Rat Ghrelin (aa 252-267)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Ghrl
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.