Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Growth Hormone ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349744 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349744 Supplier Novus Biologicals™ Supplier No. NBP349744
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Rat Growth Hormone in samples. No significant cross-reactivity or interference between Rat Growth Hormone and analogues was observed
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 2688
Gene Symbol GH1
Quantity 1 Kit
Research Discipline Biologically Active Proteins
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1 Pituitary growth hormone, hGH-N
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.