Learn More
Sino Biological Rat Her2/ERBB2 Protein 50µg
Supplier:  Sino Biological 80079RCCH150
Encompass_Preferred
A DNA sequence encoding the Rat ErbB2 (P06494) extracellular domain (Met1-Thr654) was expressed and purified, with seven additional amino acids (ALEVLFQ) at the C-terminus.
Catalog No. 50-000-84081
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.