Learn More
Description
Specifications
Specifications
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 37.5 pg/mL |
| Product Line | Novus |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat Hsp-60 in samples. No significant cross-reactivity or interference between Rat Hsp-60 and analogues was observed. |
| Gene ID (Entrez) | 3329 |
| Gene Symbol | HSPD1 |
| Host Species | Rat |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
