Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat IFN-gamma Quantikine ELISA Kit
SDP

Catalog No. SRIF00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SRIF00 6 Plates
RIF00 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SRIF00 Supplier R&D Systems™ Supplier No. SRIF00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat IFN-gamma Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat IFN-gamma Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat IFN-gamma.

Specifications

Assay Range 31.20 to 2000 pg/mL
Assay Sensitivity 10 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Rat IFN-gamma
Sufficient For 6 x 96-well Plates
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 25712
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50 μL), Serum (50 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.