Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Ifng (P01581) Recombinant Protein

Catalog No. 89948400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-400 Supplier Abnova™ Supplier No. P5152
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: QGTLIESLESLKNYFNSSSMDAMEGKSLLLDIWRNWQKDGNTKILESQIISFYLRLFEVLKDNQAISNNISVIESHLITNFFSNSKAKKDAFMSIAKFEVNNPQIQHKAVNELIRVIHQLSPESSLRKRKRSRC

Specifications

Accession Number P01581
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 25712
Molecular Weight (g/mol) 16kDa
Name Ifng (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 100 μg
Immunogen QGTLIESLESLKNYFNSSSMDAMEGKSLLLDIWRNWQKDGNTKILESQIISFYLRLFEVLKDNQAISNNISVIESHLITNFFSNSKAKKDAFMSIAKFEVNNPQIQHKAVNELIRVIHQLSPESSLRKRKRSRC
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1EU/μg) (determined by LAL gel clot method)
Gene Alias IFNG2
Common Name Ifng
Gene Symbol Ifng
Biological Activity The ED(50) was determined by the cytopathic inhibition assay with murine L929 cells infected by EMC virus, and was found to be < 1ng/mL, corresponding to a specific activity of > 1 x 106 units/mg.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.