Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Ig2A, Alexa Fluor 594, Isotype Control, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems IC006T
This item is not returnable.
View return policy
Description
IgG2a Monoclonal specifically detects IgG2a in Rat samples. It is validated for Control.Specifications
IgG2a | |
54447 | |
Control 5 uL/10^6 cells | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Selected for use as rat IgG 2A isotype control for flow cytometry applications. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Monoclonal | |
Alexa Fluor 594 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
KLH | |
200 Tests | |
Secondary | |
Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction