Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. HAF005 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAF005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HAF005 Supplier R&D Systems Supplier No. HAF005

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for ELISA∣Immunohistochemistry∣Simple Western∣Western Blot.

Specifications

Antigen IgG
Applications ELISA, Immunohistochemistry, Simple Western, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen Rat IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects rat IgG 1 IgG 2A IgG 2B heavy and light chains in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with human IgG and rabbit IgG is observed.
Target Species Rat
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.