Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgG Isotype Control

Catalog No. PI31933
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31933 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31933 Supplier Invitrogen™ Supplier No. 31933
Add to Cart
Edge
Add to Cart

Rat Polyclonal Isotype Control

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against goat anti-rat whole serum is the same as that against goat anti-rat IgG, Fc fragment specific. No precipitin line was detected against goat anti-rat IgA, alpha chain specific or goat anti-rat IgM, μ chain specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: Store at 2-8°C under sterile conditions. Dilute only enough antibody for a single day's use. Country of Origin: USA.

Rat IgG is a type of immunoglobulin G (IgG) antibody that is commonly used in scientific research and diagnostics. It is produced by rats in response to foreign antigens, and is widely used as a tool for detecting and measuring the presence of specific proteins or other molecules in biological samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG
Applications Inhibition Assays, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 11.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.6
Gene Alias Immunoglobulin G; ImmunoglobulinG
Host Species Rat
Purification Method Purified
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C
Product Type Isotype Control
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.