Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Catalog No. VC005025
Change view
Click to view available options
Quantity:
125 mL
25 mL
250 mL (2 x 125 mL)
50 mL (2 x 25 mL)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VC005025 25 mL
VC005050 50 mL (2 x 25 mL)
VC005125 125 mL
VC005250 250 mL (2 x 125 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. VC005025 Supplier R&D Systems Supplier No. VC005025
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Goat
Immunogen Rat IgG
Purification Method Affinity Purified
Quantity 25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Rat
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.