Learn More
Description
The A110-1 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, or rat cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone A110-1 was selected as an isotype control following screening for low background on a variety of mouse and rat tissues.
Host Species: Rat
Clone: A110-1
Isotype: IgG1, λ
Immunogen: Trinitrophenal-KLH
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | A110-1 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 421 |
| Host Species | Rat |
| Immunogen | Trinitrophenol-KLH |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Content And Storage | 4°C. Store in dark. |
| Isotype | IgG1 λ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.