Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG1 Isotype Control, R&D Systems™

Catalog No. MAB005
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB005 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB005 Supplier R&D Systems Supplier No. MAB005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 49 publications

IgG1 Monoclonal specifically detects IgG1 in Rat samples. It is validated for Control.

Specifications

Antigen IgG1
Classification Monoclonal
Clone 43414
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Gene Symbols IGHG1
Host Species Rat
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Test Specificity Selected for use as rat IgG 1 isotype control. In direct ELISAs, no cross-react with recombinant human (rh) 6Ckine, recombinant mouse (rm) 6Ckine, rhACT II (rhMIP-1 beta ), rhBLC/BCA-1, rmC10, recombinant rat (rr) CINC-1, rrCINC-2 alpha , rrCINC-2 beta , rmCRG-2, rhENA-78, rhEotaxin, rmEotaxin, rhEotaxin 2, rhFractalkine, rmFractalkine, rhGCP-2, rmGCP-2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhHCC-1, rhHCC-4, rhI-309, rhIL-8, rhIL-16, rhIP-10, rmJE, rmKC, rmLymphotactin, rmMARC, rhMCP-1, rhMCP-1 R, rhMCP-2, rhMCP-3, rhMCP-4, rmMCP-5, rhMDC, rmMDC, rhMIG, rmMIG, rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rmMIP-1 gamma , rhMIP-1 delta , rmMIP-2, rhMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , rmMIP-3 beta , recombinant viral (rv) MIP-I, rvMIP-II, rvMIP-III, rhMPIF-1, rhNAP-2, rhParc, rhRANTES, rmRANTES, rhSDF-1 alpha , rmSDF-1 alpha , rhSDF-1 beta , rhTarc, rmTCA-3, rhTeck, or rmTeck.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.