Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rat IgG2a, FITC, Clone: 2A3, Negative Control, EMD Millpore™
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1781
Description
Specifically detects IgG2a Clone: 2A3 in samples, and it is validated for Flow Cytometry
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Specifications
IgG2a | |
Monoclonal | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
Flow Cytometry | |
2A3 | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography. | |
Proprietary immunogen | |
100 μg | |
Inflammation, Immunology | |
Rat IgG2a | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction