Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgG2a Isotype Control, Biotin

Catalog No. PIPA533213
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533213 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533213 Supplier Invitrogen™ Supplier No. PA533213
Add to Cart
Add to Cart

Rat Monoclonal Isotype Control

PA5-33213 has been shown to be negative on all mouse cells and cell lines tested. This antibody recognizes a human cell surface marker, and therefore cannot be used as a negative control in this species Test results have shown that PA5-33213 is also suitable for use as a negative control with canine tissues. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2a
Applications Control, Flow Cytometry
Classification Monoclonal
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Rat
Immunogen Human lymphocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Isotype Control
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.