Learn More
Invitrogen™ Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™, Invitrogen™
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: APC-eFluor™ 780 Rat IgG2a Isotype Control (APC-Alexa Fluor™ 750 replacement) has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2a K Isotype Control APC-eFluor™ 780 has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | Rat IgG2a kappa |
| Applications | Control, Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBR2a |
| Concentration | 0.2 mg/mL |
| Conjugate | APC-eFluor 780 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 |
| Host Species | Rat |
| Purification Method | Affinity chromatography |
| Show More |
Frequently Asked Questions (FAQs)
Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.