Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgG2a kappa Isotype Control (eBR2a), Biotin, eBioscience™, Invitrogen™

Catalog No. 5011857
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-118-57 500 μg
50-112-9598 50 μg
50-112-9638 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-118-57 Supplier Invitrogen™ Supplier No. 13432185
Add to Cart
Add to Cart

Rat Isotype Control

Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in immunocytochemistry, immunohistochemistry, flow cytometric analysis, and ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and should be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2a kappa
Applications Control, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone eBR2a
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.