Learn More
Rat IgG2B Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems IC013N
This item is not returnable.
View return policy
Description
IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for Control.Specifications
IgG2b | |
141945 | |
Control 5 uL/10^6 cells | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Selected for use as a rat IgG 2B isotype control for flow cytometry applications. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
KLH | |
200 Tests | |
Rat | |
IgG2b |