Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2B Isotype Control, R&D Systems™

Catalog No. MAB0061
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB0061 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB0061 Supplier R&D Systems Supplier No. MAB0061
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 37 publications

IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for Control.

Specifications

Antigen IgG2b
Classification Monoclonal
Clone 141945
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Host Species Rat
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Test Specificity Selected for use as a rat IgG 2B isotype control. In direct ELISAs, does not cross-react with 102 different cytokines.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.