Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat IgG2b kappa Isotype Control (eB149/10H5), FITC, eBioscience™, Invitrogen™

Catalog No. 509805
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-980-5 500 μg
50-112-9618 50 μg
50-112-9627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-980-5 Supplier Invitrogen™ Supplier No. 11403185
Add to Cart
Edge
Add to Cart

Rat Isotype Control

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2b, kappa, isotype control has been reported for use as a control for immunofluorescent staining, flow cytometric analysis, immunohistochemistry and immunocytochemistry. Applications Tested: This rat IgG2b, kappa, isotype control has been tested by immunofluorescent staining and flow cytometric analysis of mouse splenocyte suspensions and can be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2b kappa
Applications Control, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone eB149/10H5
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.