Learn More
Invitrogen™ Rat IgG2b kappa Isotype Control (eB149/10H5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Isotype Control
Supplier: Invitrogen™ 46403180

Description
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: PerCP-eFluor® 710 Rat IgG2b Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: PerCP-eFluor® 710 Rat IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells.Use at the same concentration as experimental antibody. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.
Specifications
Rat IgG2b kappa | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Control, Flow Cytometry | |
eB149/10H5 | |
PerCP-eFluor 710 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Isotype Control | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.