Learn More
Invitrogen™ Rat IgM Isotype Control (eBRM), PE, eBioscience™, Invitrogen™
Description
Description: The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control PE has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | Rat IgM |
| Applications | Control, Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBRM |
| Concentration | 0.2 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202 |
| Host Species | Rat |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.