Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit
SDP

Catalog No. SRLB00 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
6 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SRLB00 6 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SRLB00 Supplier R&D Systems™ Supplier No. SRLB00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat IL-1 beta/IL-1F2.

Specifications

Assay Range 31.20 to 2000 pg/mL
Assay Sensitivity 5 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Rat IL-1 beta
Sufficient For 6 x 96-well Plates
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 24494
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50 μL), Serum (17 μL), EDTA Plasma (17 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL-1 b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.