Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat IL-10 DuoSet ELISA
SDP

Catalog No. DY522 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY522 15 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY522 Supplier R&D Systems™ Supplier No. DY522
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rat IL-10.

Specifications

Assay Range 62.50 - 4000 pg/mL
Conjugate Biotin
Quantity 15 96-well Plates
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.