Learn More
R&D Systems™ Rat IL-18 BPa (NP_445826) VersaClone cDNA
Description
- Il18bp interleukin 18 binding protein [ Rattus norvegicus (Norway rat) ]
- The IL-18 BPa/IL18BP insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- Interleukin 18 binding proteins are secreted glycoproteins which antagonize IL-18 activity by binding IL-18 and blocking its biological activity.
- Alternative splicing of the IL-18 BP gene produces at least four isoforms that differ in affinity for IL-18.
- The IL-18 BP isoforms a and c each contain one immunoglobulin (Ig)-like C2-type domain while isoforms b and d lack a complete Ig-like domain.
- IL-18 BPa functions as an inhibitor of the proinflammatory cytokine, IL18.
- It binds IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production, resulting in reduced Thelper type 1 immune responses.
- It is constitutively expressed and secreted in mononuclear cells.
- Elevated level of IL-18 BPa is detected in the intestinal tissues of patients with Crohn's disease.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | rIl18bp |
| Accession Number | NP_445826 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 601 bp |
| Gene ID (Entrez) | 10068 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rat |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.