Learn More
ACROBiosystems ACROBiosystems Rat IL-2 R beta&IL-2 R gamma Heterodimer Protein, His Tag&Twin-Strep Tag (MALS verified)
Supplier:  ACROBiosystems ILGR5283100UG
Encompass_Preferred
Recombinant Protein;100UG;Rat IL-2RB&IL-2RG Heterodimer Protein, His Tag&Twin-Strep Tag (ILG-R5283) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 239 (IL-2RB) & Ser 25 - Ala 262 (IL-2RG) (Accession # P26896-1 (IL-2RB) & Q68FU6-1 (IL-2RG)).;Rat IL-2RB&IL-2RG Heterodimer Protein, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RG, has a calculated MW of 30.2 kDa (IL-2RB) and 34.7 kDa (IL-2RG). Subunit IL-2RB is fused with a polyhistidine tag at the C-terminus and subunit IL-2RG is fused with Twin-Strep tag at the C-terminus. The reducing (R) protein migrates as 40-50 kDa and 55-65 kDa respectively due to glycosylation.
Catalog No. 50-210-7107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.