Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat IL-21 Recombinant Protein, PeproTech®

Catalog No. 40041250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40041250UG 250 μg
40041100UG 100 μg
400-41-10UG 10 μg
400-41-1MG 1 mg
400-41-2UG 2 μg
40041500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 40041250UG Supplier Gibco™ Supplier No. 40041250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat IL-21 is a 15.3 kDa protein containing 130 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A3QPB9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 365769
Molecular Weight (g/mol) 15.3 kDa
Name Rat IL-21
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CVID11; C x 2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11
Common Name IL-21
Gene Symbol IL21
Biological Activity Determined by its ability to stimulate the proliferation of human ANBL-6 cells. The expected ED50 is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MHKSSPQRPD HLLIRLRHLM DIVEQLKIYE NDLDPELLTA PQDVKGQCEH EAFACFQKAK LKPSNTGNNK TFINDLLAQL RRRLPAKRTG NKQRHMAKCP SCDLYEKKTP KEFLERLKWL LQKMIHQHLS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.