Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat IL-5 Recombinant Protein, PeproTech®

Catalog No. 4000510UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2.5 x 200 μg
200 μg
5 x 200 μg
50 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400-05-10UG 10 μg
40005100UG 100 μg
40-005-01M 5 x 200 μg
40-005-500 2.5 x 200 μg
40005200UG 200 μg
400-05-2UG 2 μg
400-05-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 400-05-10UG Supplier Gibco™ Supplier No. 4000510UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

400-05-1MG will be provided as 5 x 200 μg (400-05-200UG). 400-05-500UG will be provided as multiple 100 μg (400-05-100UG) and 200 μg (400-05-200UG) vials. Recombinant Rat IL-5 is a 26.0 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08125
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 24497
Molecular Weight (g/mol) 26 kDa
Name Rat IL-5
Purification Method Purified
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Common Name IL-5
Gene Symbol Il5
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MEIPMSTVVK ETLIQLSTHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEILFQNL SLIKKYIDGQ KEKCGEERRK TRHFLDYLQE FLGVMSTEWA MEV
Content And Storage -20°C
Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.