Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat IL-6 Quantikine ELISA Kit
SDP

Catalog No. R6000B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6000B 1 Plate
SR6000B 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R6000B Supplier R&D Systems™ Supplier No. R6000B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat IL-6 Quantikine ELISA Kit quantifies rat IL-6.

Specifications

Assay Range 62.50 to 4000 pg/mL
Assay Sensitivity 36 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Rat IL-6
Sufficient For 1 x 96-well Plate
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Gene Symbol Il6
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50 μL), Serum (50 μL), EDTA Plasma (50 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.