Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat IL-6 Recombinant Protein, PeproTech®

Catalog No. 400062UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400-06-2UG 2 μg
40006100UG 100 μg
400-06-10UG 10 μg
40-006-01M 2 x 500 μg
40006500UG 500 μg
400-06-50UG 50 μg
40006250U 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 400-06-2UG Supplier Gibco™ Supplier No. 400062UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

400-06-1MG will be provided as 2 x 500 μg (400-06-500UG). Recombinant Rat IL-6 is a 21.7 kDa protein containing 188 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20607
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 24498
Molecular Weight (g/mol) 21.7 kDa
Name Rat IL-6
Purification Method Purified
Quantity 2 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6
Common Name IL-6
Gene Symbol Il6
Biological Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of IL-6-dependent murine 7 TD1 cells is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT
Content And Storage -20°C
Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of IL-6-dependent murine 7TD1 cells is ≤ 0.01 ng/ml, corresponding to a specific activity of ≥ 1 x 10^8 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.