Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat IL-9 Recombinant Protein, PeproTech®

Catalog No. 400182UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400-18-2UG 2 μg
40018100UG 100 μg
400-18-10UG 10 μg
400-18-1MG 1 mg
40018250UG 250 μg
40018500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 400-18-2UG Supplier Gibco™ Supplier No. 400182UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat IL-9 is a 14.3 kDa protein of 127 amino acid residues, including 10 cysteine residues that are fully conserved between the human and murine proteins. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 116558
Molecular Weight (g/mol) 14.3 kDa
Name Rat IL-9
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40
Common Name IL-9
Gene Symbol Il9
Biological Activity Measured by its ability to enhance MC/9 cell proliferation.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQRCSTSWGI QHTSYLIENL KDDPSSKCSC SANVTSCLCL PIPSDDCTTP CFQEGMSQVT NATQQSKFSP FFFRVKRIVE TLKSNKCQFF SCEKPCNQTT AGNTVSFLKS LLKTFQKTEV QVQRSRA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.